3,5-Difluorsalicylsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3,5-Difluoro-2-hydroxybenzoic acid

H32940.03
H32940.03EA 95.2 EUR
H32940.03
3,5-Difluorsalicylsäure ≥98%
3,5-Difluorsalicylsäure

Purity: 98+% CAS No.: 84376-20-5 Molecular Formula: C7H4F2O3 Formula Weight: 174.10 Melting Point: 187-191° MDL No.: MFCD06203609

Formel: C₇H₄F₂O₃
Molecular Weight: 174.10 g/mol
Schmelzpunkt: 187…191 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06203609
CAS-Nummer: 84376-20-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR