3,5-Bis(trifluormethyl)phenylhydrazin Hydrochlorid ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
H32975.06
H32975.06EA
70.3
EUR
H32975.06
3,5-Bis(trifluormethyl)phenylhydrazin Hydrochlorid ≥98%
3,5-Bis(trifluormethyl)phenylhydrazin Hydrochlorid
Purity: 98%
CAS No.: 502496-23-3
Molecular Formula: C8H6F6N2.HCl
Formula Weight: 280.60
Melting Point: ca 208° dec.
MDL No.: MFCD03094165
Formel:
C₈H₇ClF₆N₂ Molecular Weight: 280,60 g/mol Schmelzpunkt: ∼ 208 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094165 CAS-Nummer: 502496-23-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...