2-Fluor-6-methoxypyridin ≥97%

Lieferant: Thermo Scientific
Achtung

H32994.03
H32994.03EA 127 EUR
H32994.03
2-Fluor-6-methoxypyridin ≥97%
2-Fluoro-6-methoxypyridine

Purity: 97% CAS No.: 116241-61-3 Molecular Formula: C6H6FNO Formula Weight: 127.12 UN No.: 1993 MDL No.: MFCD09998161

Formel: C₆H₆FNO
Lagertemperatur: Raumtemperatur
MDL: MFCD09998161
CAS-Nummer: 116241-61-3
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR