5-(4-Methylphenyl)thiazolo[2,3-c]-1,2,4-triazole-3-thiol 96%
Lieferant: Thermo Scientific
|
Achtung
|
H33012.ME
H33012.MEEA
103
EUR
H33012.ME
5-(4-Methylphenyl)thiazolo[2,3-c]-1,2,4-triazole-3-thiol 96%
5-(4-Methylphenyl)thiazolo[2,3-c]-1,2,4-triazole-3-thiol
Purity: 96%
CAS No.: 500112-74-3
Molecular Formula: C11H9N3S2
Formula Weight: 247.34
Melting Point: 194-195°
MDL No.: MFCD06254379
Formel:
C₁₁H₉N₃S₂ Molecular Weight: 247,34 g/mol Schmelzpunkt: 194…195 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06254379 CAS-Nummer: 500112-74-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...