5-Phenyl-1,3,4-thiadiazole-2-thiol 96%
Lieferant: Thermo Scientific
|
Achtung
|
H33061.ME
H33061.MEEA
107
EUR
H33061.ME
5-Phenyl-1,3,4-thiadiazole-2-thiol 96%
5-Phenyl-1,3,4-thiadiazole-2-thiol
Purity: 96%
CAS No.: 5585-19-3
Molecular Formula: C8H6N2S2
Formula Weight: 194.28
Melting Point: 213-215°
MDL No.: MFCD01681863
Formel:
C₈H₆N₂S₂ Molecular Weight: 194,28 g/mol Schmelzpunkt: 213…215 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01681863 CAS-Nummer: 5585-19-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...