3-(4-Chloro-3-methylphenyl)propionic acid 96%

Lieferant: Thermo Fisher Scientific
Achtung

H33072.ME
H33072.MEEA 142 EUR
H33072.ME
3-(4-Chloro-3-methylphenyl)propionic acid 96%
3-(4-Chloro-3-methylphenyl)propionic acid

Purity: 96% CAS No.: 1086386-05-1 Molecular Formula: C10H11ClO2 Formula Weight: 198.65 Melting Point: 88-92° MDL No.: MFCD11655590

Formel: C₁₀H₁₁ClO₂
Schmelzpunkt: 88…92 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11655590
CAS-Nummer: 1086386-05-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR