2,1,3-Benzothiadiazol-5-carbonsäure ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H33095.MD
H33095.03
H33095.06
H33095.MDEA
60.6
EUR
H33095.MD
H33095.03
H33095.06
2,1,3-Benzothiadiazol-5-carbonsäure ≥97%
2,1,3-Benzothiadiazole-5-carboxylic acid
Formel:
C₇H₄N₂O₂S Schmelzpunkt: 228…230 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01647555 CAS-Nummer: 16405-98-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...