2-Fluor-4-(trifluormethyl)pyridin ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

H33198.06
H33198.06EA 96.5 EUR
H33198.06
2-Fluor-4-(trifluormethyl)pyridin ≥97%
2-Fluoro-4-(trifluoromethyl)pyridine

Purity: 97% CAS No.: 118078-66-3 Molecular Formula: C6H3F4N Formula Weight: 165.09 Boiling Point: 110° MDL No.: MFCD09879262

Formel: C₆H₃F₄N
Molecular Weight: 165.09 g/mol
Siedepunkt: 113 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09879262
CAS-Nummer: 118078-66-3
UN: 1992
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR