Methyl-4-amino-3-brom-5-nitrobenzoat 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Methyl 4-amino-3-bromo-5-nitrobenzoate

H33216.06
H33216.06EA 202 EUR
H33216.06
Methyl-4-amino-3-brom-5-nitrobenzoat 97%
Methyl-4-amino-3-brom-5-nitrobenzoat
Formel: C₈H₇BrN₂O₄
Molecular Weight: 275,06 g/mol
Schmelzpunkt: 107…109 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08443938
CAS-Nummer: 105655-17-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR