5-Chloro-2-(hydroxymethyl)furo[3,2-b]pyridine 97%
Lieferant: Thermo Scientific
Achtung
H33292.03
H33292.03EA 179 EUR
H33292.03
5-Chloro-2-(hydroxymethyl)furo[3,2-b]pyridine 97%
5-Chloro-2-(hydroxymethyl)furo[3,2-b]pyridine
Purity: 97% Molecular Formula: C8H6ClNO2 Formula Weight: 183.59 Melting Point: 108-110° MDL No.: MFCD20040113