5-Chloro-2-(hydroxymethyl)furo[3,2-b]pyridine 97%
Lieferant: Thermo Scientific
|
Achtung
|
H33292.03
H33292.03EA
179
EUR
H33292.03
5-Chloro-2-(hydroxymethyl)furo[3,2-b]pyridine 97%
5-Chloro-2-(hydroxymethyl)furo[3,2-b]pyridine
Purity: 97%
Molecular Formula: C8H6ClNO2
Formula Weight: 183.59
Melting Point: 108-110°
MDL No.: MFCD20040113