2-Amino-3,3,3-trifluoropropionic acid ethylester hydrochloride 96%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Ethyl 2-amino-3,3,3-trifluoropropionate hydrochloride, Ethyl 2-amino-3,3,3-trifluoropropanoate hydrochloride
H33319.03
H33319.03EA
105
EUR
H33319.03
2-Amino-3,3,3-trifluoropropionic acid ethylester hydrochloride 96%
2-Amino-3,3,3-trifluoropropionic acid ethylester hydrochloride
Purity: 96%
CAS No.: 193140-71-5
Molecular Formula: C5H8F3NO2.HCl
Formula Weight: 207.58
Melting Point: ca 149°dec.
MDL No.: MFCD14525479
Formel:
C₅H₉ClF₃NO₂ Molecular Weight: 207,58 g/mol Schmelzpunkt: ∼ 149 °C, (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD14525479 CAS-Nummer: 193140-71-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...