Coumarin 334 96%

Lieferant: Thermo Fisher Scientific
Gefahr

H33332.ME
H33332.MEEA 71.9 EUR
H33332.ME
Coumarin 334 96%
Coumarin 334

Purity: 96% CAS No.: 55804-67-6 Molecular Formula: C25H24N2O4S Formula Weight: 448.53 Melting Point: 176-181° UN No.: UN2811 MDL No.: MFCD00051336

Formel: C₁₇H₁₇NO₃
Schmelzpunkt: 176…181 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051336
CAS-Nummer: 55804-67-6
EINECS: 259-826-7
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR