3-Amino-5-brom-1H-indazol 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-Bromo-1H-indazol-3-amine

H33351.03
H33351.03EA 45.4 EUR
H33351.03
3-Amino-5-brom-1H-indazol 97%
3-Amino-5-bromo-1H-indazole
Formel: C₇H₆BrN₃
Molecular Weight: 212,05 g/mol
Schmelzpunkt: 171…174 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03426696
CAS-Nummer: 61272-71-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR