3-Brom-4-fluor-5-iodbenzaldehyd ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
H33441.03
H33441.03EA
132
EUR
H33441.03
3-Brom-4-fluor-5-iodbenzaldehyd ≥97%
3-Bromo-4-fluoro-5-iodobenzaldehyde
Formel:
C₇H₃BrFIO Molecular Weight: 328,90 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD20486165 CAS-Nummer: 1356113-37-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...