4-Bromo-1-methyl-1H-pyrazole-3-carbaldehyde 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Bromo-1-methyl-1H-pyrazole-3-carboxaldehyde
H33484.06
H33484.06EA
254
EUR
H33484.06
4-Bromo-1-methyl-1H-pyrazole-3-carbaldehyde 97%
4-Bromo-1-methyl-1H-pyrazole-3-carbaldehyde
Purity: 97%
CAS No.: 287917-96-8
Molecular Formula: C5H5BrN2O
Formula Weight: 189.01
Melting Point: 69-71°
MDL No.: MFCD00103235
Formel:
C₅H₅BrN₂O Molecular Weight: 189,02 g/mol Schmelzpunkt: 69…71 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00103235 CAS-Nummer: 287917-96-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...