4-Nitrophthalonitril ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

H33546.03 H33546.09
H33546.03EA 15.5 EUR
H33546.03 H33546.09
4-Nitrophthalonitril ≥97%
4-Nitrophthalonitril

Purity: 97% CAS No.: 31643-49-9 Molecular Formula: C8H3N3O2 Formula Weight: 173.13 Melting Point: 136-137° MDL No.: MFCD00040301

Formel: C₈H₃N₃O₂
Schmelzpunkt: 136…137 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00040301
CAS-Nummer: 31643-49-9
EINECS: 250-748-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR