2,4,6-Triphenoxy-1,3,5-triazin 96%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
H33572.18
H33572.18EA
142
EUR
H33572.18
2,4,6-Triphenoxy-1,3,5-triazin 96%
2,4,6-Triphenoxy-1,3,5-triazine
Purity: 96%
CAS No.: 1919-48-8
Molecular Formula: C21H15N3O3
Formula Weight: 357.36
Melting Point: 235-236°
MDL No.: MFCD00006048
Formel:
C₂₁H₁₅N₃O₃ Schmelzpunkt: 235…236 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006048 CAS-Nummer: 1919-48-8 EINECS: 217-644-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...