3-(3-Chloro-4-fluorophenyl)propionic acid 96%

Lieferant: Thermo Fisher Scientific
Achtung

H33592.03 H33592.06
H33592.03EA 55.7 EUR
H33592.03 H33592.06
3-(3-Chloro-4-fluorophenyl)propionic acid 96%
3-(3-Chloro-4-fluorophenyl)propionic acid

Purity: 96% CAS No.: 881190-93-8 Molecular Formula: C9H8ClFO2 Formula Weight: 202.61 Melting Point: 56-59° MDL No.: MFCD04116056

Formel: C₉H₈ClFO₂
Schmelzpunkt: 56…59 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04116056
CAS-Nummer: 881190-93-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR