5-Nitrobenzo[d]thiazole 96%
Lieferant: Thermo Scientific
|
Achtung
|
H33594.03
H33594.03EA
66.9
EUR
H33594.03
5-Nitrobenzo[d]thiazole 96%
5-Nitrobenzo[d]thiazole
Purity: 96%
CAS No.: 2942-07-6
Molecular Formula: C7H4N2O2S
Formula Weight: 180.18
Melting Point: 157-162°
MDL No.: MFCD00085891
Formel:
C7H4N2O2S Molecular Weight: 180,18 g/mol Schmelzpunkt: 157…162 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00085891 CAS-Nummer: 2942-07-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...