(R)-(-)-4-Fluorostyrene oxide ≥98%, ee 98%
Lieferant: Thermo Scientific
|
Gefahr
|
H33603.06
H33603.06EA
395
EUR
H33603.06
(R)-(-)-4-Fluorostyrene oxide ≥98%, ee 98%
(R)-(-)-4-Fluorostyrene oxide
Purity: 98+%
CAS No.: 134356-73-3
Molecular Formula: C8H7FO
Formula Weight: 138.14
Refractive Index: 1.5087
Boiling Point: 92/14mm
UN No.: UN1992
MDL No.: MFCD03788740
Formel:
C₈H₇FO Molecular Weight: 138,14 g/mol Siedepunkt: 92 °C (14 mmHg) Dichte: 1,17 Lagertemperatur: Raumtemperatur |
MDL:
MFCD03788740 CAS-Nummer: 134356-73-3 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...