3-Fluor-5-(pentafluorthio)anilin ≥97%
Lieferant: Thermo Fisher Scientific
H33680.MD
H33680.03
H33680.MDEA
146
EUR
H33680.MD
H33680.03
3-Fluor-5-(pentafluorthio)anilin ≥97%
3-Fluor-5-(pentafluorthio)anilin
Purity: 97%
Molecular Formula: C6H5F6NS
Formula Weight: 237.16
UN No.: 2811
MDL No.: MFCD16652430
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...