2-Brom-4-chlorpyridin ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
H33698.03
H33698.06
H33698.03EA
36.5
EUR
H33698.03
H33698.06
2-Brom-4-chlorpyridin ≥97%
2-Bromo-4-chloropyridine
Purity: 97%
CAS No.: 22918-01-0
Molecular Formula: C5H3BrClN
Formula Weight: 192.44
Melting Point:
Formel:
C₅H₃BrClN Molecular Weight: 192.44 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03840757 CAS-Nummer: 22918-01-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...