2-(Trifluoromethyl)quinoline 97%

Lieferant: Thermo Fisher Scientific
Achtung

H33740.MD
H33740.MDEA 64.2 EUR
H33740.MD
2-(Trifluoromethyl)quinoline 97%
2-(Trifluoromethyl)quinoline

Purity: 97% CAS No.: 347-42-2 Molecular Formula: C10H6F3N Formula Weight: 197.16 Melting Point: 59-62° MDL No.: MFCD01862002

Formel: C₁₀H₆F₃N
Schmelzpunkt: 59…62 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01862002
CAS-Nummer: 347-42-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR