3-(Trifluormethyl)benzoesäureanhydrid ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

H33779.03 H33779.06
H33779.03EA 30 EUR
H33779.03 H33779.06
3-(Trifluormethyl)benzoesäureanhydrid ≥97%
3-(Trifluoromethyl)benzoic anhydride

Purity: 97% CAS No.: 25753-15-5 Molecular Formula: C16H8F6O3 Formula Weight: 362.23 Melting Point: 74-78° MDL No.: MFCD09757594

Formel: C₁₆H₈F₆O₃
Schmelzpunkt: 74…78 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09757594
CAS-Nummer: 25753-15-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR