2-Methyl-5-(trifluormethoxy)benzylamin 96%

Lieferant: Thermo Scientific
Gefahr

H33791.03
H33791.03EA 116 EUR
H33791.03
2-Methyl-5-(trifluormethoxy)benzylamin 96%
2-Methyl-5-(trifluormethoxy)benzylamin
Formel: C₉H₁₀F₃NO
Molecular Weight: 205,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22201056
CAS-Nummer: 1373920-87-6
UN: 3267
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR