3-Amino-2-brom-5-fluorpyridin 97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2-Brom-5-fluorpyridin-3-amin, 2-Brom-5-fluor-3-pyridylamin

H33811.06
H33811.06EA 240 EUR
H33811.06
3-Amino-2-brom-5-fluorpyridin 97%
3-Amino-2-brom-5-fluorpyridin

Purity: 97% CAS No.: 884495-03-8 Molecular Formula: C5H4BrFN2 Formula Weight: 191.00 Melting Point: 97-101° MDL No.: MFCD05662413

Formel: C₅H₄BrFN₂
Molecular Weight: 191 g/mol
Schmelzpunkt: 97…101 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05662413
CAS-Nummer: 884495-03-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR