Ethyl-2-amino-4,4,4-trifluoro-3-(trifluoromethyl)butyrate 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: Ethyl 4,4,4,4',4',4'-hexafluoro-DL-valinate, 4,4,4,4',4',4'-Hexafluorovaline, ethyl ester

H33837.03
H33837.03EA 142 EUR
H33837.03
Ethyl-2-amino-4,4,4-trifluoro-3-(trifluoromethyl)butyrate 95%
Ethyl-2-amino-4,4,4-trifluoro-3-(trifluoromethyl)butyrate

Purity: 95% CAS No.: 78185-92-9 Molecular Formula: C7H9F6NO2 Formula Weight: 253.14 Refractive Index: 1.3600 Boiling Point: 70-72°/10mm MDL No.: MFCD00040845

Formel: C₇H₉F₆NO₂
Molecular Weight: 253,15 g/mol
Siedepunkt: 70…72 °C (10 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD00040845
CAS-Nummer: 78185-92-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR