3-(5-Bromo-2-methoxyphenyl)propionic acid 96%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-(5-Bromo-2-methoxyphenyl)propanoic acid

H33961.06
H33961.06EA 120 EUR
H33961.06
3-(5-Bromo-2-methoxyphenyl)propionic acid 96%
3-(5-Bromo-2-methoxyphenyl)propionic acid

Purity: 96% CAS No.: 82547-30-6 Molecular Formula: C10H11BrO3 Formula Weight: 259.10 Melting Point: 114-116° MDL No.: MFCD09258907

Formel: C₁₀H₁₁BrO₃
Schmelzpunkt: 114…116 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09258907
CAS-Nummer: 82547-30-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR