Ethyl-3-chlor-5-(trifluormethyl)picolinat 96%

Lieferant: Thermo Scientific
Achtung

Synonyme: Ethyl 3-chloro-5-(trifluoromethyl)picolinate, Ethyl 3-chloro-5-(trifluoromethyl)pyridine-2-carboxylate

H33967.09
H33967.09EA 342 EUR
H33967.09
Ethyl-3-chlor-5-(trifluormethyl)picolinat 96%
Ethyl-3-chlor-5-(trifluormethyl)picolinat
Formel: C₉H₇ClF₃NO₂
Molecular Weight: 253,60 g/mol
Siedepunkt: 64…66 °C (0,008 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD06656414
CAS-Nummer: 128073-16-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR