2-(2-Phthalimidoethoxy)acetic acid 97%

Lieferant: Thermo Fisher Scientific

H33988.03 H33988.06
H33988.03EA 52.5 EUR
H33988.03 H33988.06
2-(2-Phthalimidoethoxy)acetic acid 97%
2-(2-Phthalimidoethoxy)acetic acid

Purity: 97% CAS No.: 69676-65-9 Molecular Formula: C12H11NO5 Formula Weight: 249.22 Melting Point: 130-132° MDL No.: MFCD06661364

Formel: C₁₂H₁₁NO₅
Schmelzpunkt: 130…132 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06661364
CAS-Nummer: 69676-65-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR