6-(Trifluoromethyl)pyridine-3-sulphonyl chloride 97%

Lieferant: Thermo Scientific
Gefahr

H34033.03
H34033.03EA 95.1 EUR
H34033.03
6-(Trifluoromethyl)pyridine-3-sulphonyl chloride 97%
6-(Trifluoromethyl)pyridine-3-sulphonyl chloride

Purity: 97% CAS No.: 959996-58-8 Molecular Formula: C6H3ClF3NO2S Formula Weight: 245.61 Melting Point: 42-45° UN No.: UN3261 MDL No.: MFCD10699112

Formel: C₆H₃ClF₃NO₂S
Molecular Weight: 245,61 g/mol
Schmelzpunkt: 42…45 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD10699112
CAS-Nummer: 959996-58-8
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR