2,4-Dichlor-3-formylpyridin ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2,4-Dichloronicotinaldehyde, 2,4-Dichloropyridine-3-carboxaldehyde, 2,4-Dichloro-3-formylpyridine, 2,4-Dichloro-3-pyridinecarboxaldehyde

H34053.03
H34053.03EA 81.4 EUR
H34053.03
2,4-Dichlor-3-formylpyridin ≥97%
2,4-Dichlor-3-formylpyridin
Formel: C₆H₃Cl₂NO
Molecular Weight: 176,00 g/mol
Schmelzpunkt: 72…75 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07437909
CAS-Nummer: 134031-24-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR