3-(Trifluormethyl)cyclohexanon ≥97%

Lieferant: Thermo Scientific
Achtung

H34087.03
H34087.03EA 90.3 EUR
H34087.03
3-(Trifluormethyl)cyclohexanon ≥97%
3-(Trifluoromethyl)cyclohexanone
Formel: C₇H₉F₃O
Molecular Weight: 166,14 g/mol
Siedepunkt: 82 °C (29 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD00034662
CAS-Nummer: 585-36-4
UN: 1224
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR