2-Fluor-4-(pentafluorthio)anilin ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

H34099.MD H34099.03
H34099.MDEA 198 EUR
H34099.MD H34099.03
2-Fluor-4-(pentafluorthio)anilin ≥97%
2-Fluor-4-(pentafluorthio)anilin

Purity: 97% CAS No.: 1240257-25-3 Molecular Formula: C6H5F6NS Formula Weight: 237.16 UN No.: 2810 MDL No.: MFCD16652511

Formel: C₆H₅F₆NS
Lagertemperatur: Raumtemperatur
MDL: MFCD16652511
CAS-Nummer: 1240257-25-3
UN: 2810
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR