3-Amino-5-fluorpyridin 98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
H34111.06
H34111.06EA
202
EUR
H34111.06
3-Amino-5-fluorpyridin 98%
3-Amino-5-fluoropyridine
Formel:
C₅H₅FN₂ Schmelzpunkt: 85…89 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00234073 CAS-Nummer: 210169-05-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...