Methyl-1H-pyrazol-5-carboxylat 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Methyl 1H-pyrazole-3-carboxylate
H34148.03
H34148.03EA
101
EUR
H34148.03
Methyl-1H-pyrazol-5-carboxylat 97%
Methyl-1H-pyrazole-3-carboxylate
Purity: 97%
CAS No.: 15366-34-4
Molecular Formula: C5H6N2O2
Formula Weight: 126.11
Melting Point: 141-143°
MDL No.: MFCD00649381
Formel:
C₅H₆N₂O₂ Molecular Weight: 126.11 g/mol Schmelzpunkt: 141…143 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00649381 CAS-Nummer: 15366-34-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...