[4-(4-Chlorophenyl)phenyl]sulphonyl chloride 96%
Lieferant: Thermo Fisher Scientific
H34182.09
H34182.03
H34182.09EA
242
EUR
H34182.09
H34182.03
[4-(4-Chlorophenyl)phenyl]sulphonyl chloride 96%
Purity: 96%
CAS No.: 20443-74-7
Molecular Formula: C12H8Cl2O2S
Formula Weight: 287.17
Melting Point: 107-108°
UN No.: UN3261
MDL No.: MFCD01631918
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...