3-(2,4,6-Trimethoxyphenyl)propionsäure 96%

Lieferant: Thermo Scientific
Achtung

H34208.09
H34208.09EA 0 EUR
H34208.09
3-(2,4,6-Trimethoxyphenyl)propionsäure 96%
3-(2,4,6-Trimethoxyphenyl)propionsäure

Purity: 96% CAS No.: 74737-05-6 Molecular Formula: C12H16O5 Formula Weight: 240.26 Melting Point: 140-144° MDL No.: MFCD00045944

Formel: C₁₂H₁₆O₅
Molecular Weight: 240,26 g/mol
Schmelzpunkt: 140…144 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00045944
CAS-Nummer: 74737-05-6
EINECS: 277-978-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR