3-Amino-5-iod-1H-indazol 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H34227.03
H34227.06
H34227.03EA
60.3
EUR
H34227.03
H34227.06
3-Amino-5-iod-1H-indazol 97%
3-Amino-5-iodo-1H-indazole
Formel:
C₇H₆IN₃ Schmelzpunkt: 177…179 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09261138 CAS-Nummer: 88805-76-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...