Diethylcyclohexan-1,1-dicarboxylat 95%

Lieferant: Thermo Scientific
Achtung

H34412.MD
H34412.MDEA 0 EUR
H34412.MD
Diethylcyclohexan-1,1-dicarboxylat 95%
Diethylcyclohexan-1,1-dicarboxylat

Purity: 95% CAS No.: 1139-13-5 Molecular Formula: C12H20O4 Formula Weight: 228.29 Boiling Point: 106-108°/1mm MDL No.: MFCD02093457

Formel: C₁₂H₂₀O₄
Molecular Weight: 228,29 g/mol
Siedepunkt: 106…108 °C (1 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD02093457
CAS-Nummer: 1139-13-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR