3-(2,3-Dimethoxyphenyl)propionsäure 96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-(2,3-Dimethoxyphenyl)propanoic acid
H34491.06
H34491.06EA
0
EUR
H34491.06
3-(2,3-Dimethoxyphenyl)propionsäure 96%
3-(2,3-Dimethoxyphenyl)propionsäure
Purity: 96%
CAS No.: 10538-48-4
Molecular Formula: C11H14O4
Formula Weight: 210.23
Melting Point: 58-60°
MDL No.: MFCD00029821
Formel:
C₁₁H₁₄O₄ Schmelzpunkt: 58…60 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00029821 CAS-Nummer: 10538-48-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...