8-Brom-6-chlorimidazo[1,2-a]pyridin 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H35094.03
H35094.03EA
125
EUR
H35094.03
8-Brom-6-chlorimidazo[1,2-a]pyridin 95%
8-Brom-6-chlorimidazo[1,2-a]pyridin
Formel:
C₇H₄BrClN₂ Schmelzpunkt: 125…128 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD12828062 CAS-Nummer: 957187-27-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...