3-(Pentafluorthio)anilin ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
3-Aminophenylsulphurpentafluoride, 3-Aminophenylsulphur pentafluoride
H35190.03
H35190.05
H35190.06
H35190.03EA
66.7
EUR
H35190.03
H35190.05
H35190.06
3-(Pentafluorthio)anilin ≥97%
3-(Pentafluorthio)anilin
Formel:
C₆H₆F₅NS Molecular Weight: 218.17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00674199 CAS-Nummer: 2993-22-8 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...