3-(Pentafluorthio)anilin ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 3-Aminophenylsulphurpentafluoride, 3-Aminophenylsulphur pentafluoride

H35190.03 H35190.05 H35190.06
H35190.03EA 66.7 EUR
H35190.03 H35190.05 H35190.06
3-(Pentafluorthio)anilin ≥97%
3-(Pentafluorthio)anilin
Formel: C₆H₆F₅NS
Molecular Weight: 218.17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00674199
CAS-Nummer: 2993-22-8
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR