3-Bromimidazo[1,2-a]pyridin-6-carbonitril 95%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
H35421.MD
H35421.03
H35421.MDEA
53.5
EUR
H35421.MD
H35421.03
3-Bromimidazo[1,2-a]pyridin-6-carbonitril 95%
3-Bromimidazo[1,2-a]pyridin-6-carbonitril
Formel:
C₈H₄BrN₃ Schmelzpunkt: 207…209 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06659617 CAS-Nummer: 885950-21-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...