3-Brom-6-chlorimidazo[1,2-a]pyridin 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H35558.03
H35558.03EA
61.2
EUR
H35558.03
3-Brom-6-chlorimidazo[1,2-a]pyridin 95%
3-Brom-6-chlorimidazo[1,2-a]pyridin
Formel:
C₇H₄BrClN₂ Schmelzpunkt: 105…107 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08667858 CAS-Nummer: 886371-28-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...