Methyl-3-bromimidazo[1,2-a]pyridin-7-carboxylat 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H35994.03
H35994.03EA
121
EUR
H35994.03
Methyl-3-bromimidazo[1,2-a]pyridin-7-carboxylat 95%
Methyl-3-bromimidazo[1,2-a]pyridin-7-carboxylat
Formel:
C₉H₇BrN₂O₂ Schmelzpunkt: 156…157 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD10566741 CAS-Nummer: 342613-63-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...