1-Octanol (natürlich) 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
n-Octanol
H36188.AP
H36188.0D
H36188.0F
H36188.APEA
32.8
EUR
H36188.AP
H36188.0D
H36188.0F
1-Octanol (natürlich) 98%
1-Octanol
Purity: 98%
CAS No.: 111-87-5
Molecular Formula: C8H18O
Formula Weight: 130.23
Flash Point: 81°(178°F)
Physical Form: Liquid
Density: 0.825
Refractive Index: 1.4295
Appearance: Colourless
Odour: Sweet
Melting Point: -16°
Boiling Point: 193-195°
MDL No.: MFCD00002988
Formel:
H₃C(CH₂)₆CH₂OH Siedepunkt: 193…195 °C Schmelzpunkt: –16 °C Dichte: 0,825 Flammpunkt: 81 °C (178 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002988 CAS-Nummer: 111-87-5 EINECS: 203-917-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...