Palladium : Platinum (2.5:2.5)% auf Kohlenstoff, Paste Type 122
Lieferant: Thermo Fisher Scientific
H36548.06
H36548.14
H36548.22
H36548.06EA
93.8
EUR
H36548.06
H36548.14
H36548.22
Palladium : Platinum (2.5:2.5)% auf Kohlenstoff, Paste Type 122
Palladium : Platinum
CAS No.: 7440-05-3
Physical Form: Solid
Appearance: Black
Odour: Odourless
MDL No.: MFCD03613605
CAS No.: 7440-05-3
Physical Form: Solid
Appearance: Black
Odour: Odourless
MDL No.: MFCD03613605
Formel:
Pd:Pt Lagertemperatur: Raumtemperatur |
MDL:
MFCD03613605 CAS-Nummer: 7440-05-3 EINECS: 231-115-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...