Piperazin Hexahydrat 97+%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Diethylendiamin Hexahydrat

H37069.0B H37069.30
H37069.0BEA 112 EUR
H37069.0B H37069.30
Piperazin Hexahydrat 97+%
Piperazin Hexahydrat

Purity: 97+% CAS No.: 142-63-2 Molecular Formula: C4H10N2.xH2O Formula Weight: 86.14anhy Flash Point: 100°(212°F) Physical Form: Crystalline powder or flake Appearance: White Odour: Weak, characteristic pH: 11-12 Melting Point: 42-46° Boiling Point: 125-130° UN No.: UN2579 MDL No.: MFCD00149389

Formel: C₄H₁₀N₂·6H₂O
Siedepunkt: 125…130 °C
Schmelzpunkt: 42…46 °C
Flammpunkt: >100 °C (212 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149389
CAS-Nummer: 142-63-2
EINECS: 203-808-3
UN: 2579
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR